Please login to your online account to display your discounted pricing

anti-Eosinophil, Clone: BMK-13, Novus Biologicals™

$362.00 - $1,067.17

Specifications

Clone BMK-13
Host Species Mouse
Isotype IgG1
Immunogen The antibody reacts with the Eosinophil Major Basic Protein antigen.
Antigen Eosinophil
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP1421401M

 
novus biologicals canada
NBP1421401ML
Unlabeled 1mL Each for $362.00

NBP1421405M

 
novus biologicals canada
NBP1421405ML
Unlabeled 5mL Each for $1,067.17
Description & Specifications

Specifications

Clone BMK-13
Host Species Mouse
Isotype IgG1
Immunogen The antibody reacts with the Eosinophil Major Basic Protein antigen.
Antigen Eosinophil
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Test Specificity BMK-13 binds to the 10 kD eosinophil Major Basic Protein (MBP) of both resting and activated eosinophils in cytospins and frozen sections of bronchial and skin biopsies of allergic sites and normal sites and thus can be used as a pan-eosinophil marker. The antibody BMK-13 stains in frozen sections of bronchial biopsies from atopic asthmatics, rhinitics and normal non-atopic subjects, substantially higher counts of positive cells when compared to EG1, EG2 and chromotrope 2R. BMK-13 cross-reacts weakly with human basophils, which also contain low level of this protein. It does not cross-react with any other human protein or cell.
Formulation PBS + 0.1% BSA
Concentration 0.1mg/mL