Please login to your online account to display your discounted pricing

anti-Eosinophil, Clone: BMK-13, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1421405ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1421405M

  • / Each
 

Description & Specifications

Specifications

Quantity 5mL
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Isotype IgG1
Host Species Mouse
Cross Reactivity Human
Regulatory Status RUO
Clone BMK-13
Format Purified
Formulation PBS + 0.1% BSA
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity BMK-13 binds to the 10 kD eosinophil Major Basic Protein (MBP) of both resting and activated eosinophils in cytospins and frozen sections of bronchial and skin biopsies of allergic sites and normal sites and thus can be used as a pan-eosinophil marker. The antibody BMK-13 stains in frozen sections of bronchial biopsies from atopic asthmatics, rhinitics and normal non-atopic subjects, substantially higher counts of positive cells when compared to EG1, EG2 and chromotrope 2R. BMK-13 cross-reacts weakly with human basophils, which also contain low level of this protein. It does not cross-react with any other human protein or cell.
Concentration 0.1mg/mL
Conjugate Unlabeled
Antigen Eosinophil
Immunogen The antibody reacts with the Eosinophil Major Basic Protein antigen.