Please login to your online account to display your discounted pricing

anti-Endothelin-1, Clone: IC4, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11017251

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11017251

  • / Each
 

Description & Specifications

Specifications

Antigen Endothelin-1
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Radioimmunoassay
Regulatory Status RUO
Immunogen C-Terminal fragment
Concentration 1.0mg/mL
Primary or Secondary Primary
Test Specificity NB110-17251 recognizes endothelin ET-1 (10% cross-reactivity with ET-3). Optimal working dilutions must be determined by the end user. Not yet tested in other species.
Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1906
Host Species Mouse
Isotype IgG1
Gene Accession No. P05305
Clone IC4
Conjugate Unlabeled
Format Purified
Formulation Constituents: PBS. pH 7.2
Cross Reactivity Human