Please login to your online account to display your discounted pricing

anti-Endothelin-1, Clone: IC4, Novus Biologicals™

$219.00 - $769.00

Specifications

Clone IC4
Host Species Mouse
Isotype IgG1
Immunogen C-Terminal fragment
Antigen Endothelin-1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB110172511

 
novus biologicals canada
NB110172511MG
Unlabeled 1mg Each for $769.00

NB11017251

 
novus biologicals canada
NB11017251
Unlabeled 0.1mg Each for $219.00
Description & Specifications

Specifications

Clone IC4
Host Species Mouse
Isotype IgG1
Immunogen C-Terminal fragment
Antigen Endothelin-1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Radioimmunoassay
Test Specificity NB110-17251 recognizes endothelin ET-1 (10% cross-reactivity with ET-3). Optimal working dilutions must be determined by the end user. Not yet tested in other species.
Gene Accession No. P05305
Gene ID (Entrez) 1906
Formulation Constituents: PBS. pH 7.2
Concentration 1.0mg/mL