Please login to your online account to display your discounted pricing

anti-Endothelin-1, Clone: IC4, Novus Biologicals™

Manufacturer: novus biologicals canada  NB110172511MG

 View all options for this product


For Research Use Only

Catalog No. NB110172511


Description & Specifications


Antigen Endothelin-1
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Radioimmunoassay
Clone IC4
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation Constituents: PBS. pH 7.2
Gene Accession No. P05305
Host Species Mouse
Immunogen C-Terminal fragment
Isotype IgG1
Quantity 1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1906
Test Specificity NB110-17251 recognizes endothelin ET-1 (10% cross-reactivity with ET-3). Optimal working dilutions must be determined by the end user. Not yet tested in other species.