Please login to your online account to display your discounted pricing

anti-EMMPRIN/CD147, Clone: 8D6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP233015

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP233015

  • / Each
 

Description & Specifications

Specifications

Antigen EMMPRIN/CD147
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human T cell line Molt 13
Cross Reactivity Human
Host Species Mouse
Quantity 0.5mL
Regulatory Status RUO
Isotype IgG1
Primary or Secondary Primary
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P35613
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 682
Clone 8D6
Concentration 0.2mg/mL
Conjugate Unlabeled