Please login to your online account to display your discounted pricing

anti-EMMPRIN/CD147, Clone: 8D6, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone 8D6
Host Species Mouse
Isotype IgG1
Immunogen Human T cell line Molt 13
Antigen EMMPRIN/CD147
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP233015

 
novus biologicals canada
NBP233015
Unlabeled 0.5mL Each for $321.00

NBP2330151M

 
novus biologicals canada
NBP2330151ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone 8D6
Host Species Mouse
Isotype IgG1
Immunogen Human T cell line Molt 13
Antigen EMMPRIN/CD147
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P35613
Gene ID (Entrez) 682
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL