Please login to your online account to display your discounted pricing

anti-eIF2A, Clone: 3A7B11, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 3A7B11
Host Species Mouse
Isotype IgG1
Immunogen Partial recombinant human EIF2A (between residues 350-585) expressed in &italic_begin;E. coli&italic_end; [Uniprot Q9BY44]
Antigen eIF2A
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP226296

 
novus biologicals canada
NBP226296
Unlabeled 0.1mL Each for $309.05

NBP226296SS

 
novus biologicals canada
NBP226296SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Clone 3A7B11
Host Species Mouse
Isotype IgG1
Immunogen Partial recombinant human EIF2A (between residues 350-585) expressed in &italic_begin;E. coli&italic_end; [Uniprot Q9BY44]
Antigen eIF2A
Monoclonal or Polyclonal Monoclonal
Format Precipitation
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Western Blotting
Gene Accession No. Q9BY44
Gene ID (Entrez) 83939
Formulation PBS, 0.5% protein stabilizer
Concentration 1.0mg/mL