Please login to your online account to display your discounted pricing

anti-EGLN3/PHD3, Clone: A1.10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP122574SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP122574SS

  • / Each
 

Description & Specifications

Specifications

Antigen EGLN3/PHD3
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Clone A1.10
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation Tris-glycine, 150mM NaCl
Gene Accession No. Q9H6Z9
Host Species Murine
Immunogen Full length human PHD3/HIF Prolyl Hydroxylase 3 protein purified from Sf9 insect cells
Isotype κ
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 112399
Target Species Human