Please login to your online account to display your discounted pricing

anti-EGLN3/PHD3, Clone: A1.10, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP1-22574

 View all options for this product


For Research Use Only

Catalog No. NBP122574


Description & Specifications


Antigen EGLN3/PHD3
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Clone A1.10
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation Tris-glycine, 150mM NaCl
Gene Accession No. Q9H6Z9
Host Species Mouse
Immunogen Full length human PHD3/HIF Prolyl Hydroxylase 3 protein purified from Sf9 insect cells
Isotype κ
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 112399