Please login to your online account to display your discounted pricing

anti-EGLN3/PHD3, Clone: A1.10, Novus Biologicals™

$109.00 - $109.00

Specifications

Clone A1.10
Host Species Mouse
Isotype κ
Immunogen Full length human PHD3/HIF Prolyl Hydroxylase 3 protein purified from Sf9 insect cells
Antigen EGLN3/PHD3
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP122574SS

 
novus biologicals canada
NBP122574SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Clone A1.10
Host Species Mouse
Isotype κ
Immunogen Full length human PHD3/HIF Prolyl Hydroxylase 3 protein purified from Sf9 insect cells
Antigen EGLN3/PHD3
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Gene Accession No. Q9H6Z9
Gene ID (Entrez) 112399
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL