Please login to your online account to display your discounted pricing

anti-EGF R/ErbB1, Clone: B1D8, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP232843

 View all options for this product


For Research Use Only

Catalog No. NBP232843


Description & Specifications


Antigen EGF R/ErbB1
Applications Flow Cytometry
Applications Immunoprecipitation
Clone B1D8
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P00533
Host Species Mouse
Immunogen Microsomes from A431 cells
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1956
Test Specificity Human. Does not react with Mouse or Rat.