Please login to your online account to display your discounted pricing

anti-EGF R/ErbB1, Clone: B1D8, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone B1D8
Host Species Mouse
Isotype κ
Immunogen Microsomes from A431 cells
Antigen EGF R/ErbB1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232843

 
novus biologicals canada
NBP232843
Unlabeled 0.5mL Each for $321.00

NBP2328431M

 
novus biologicals canada
NBP2328431ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone B1D8
Host Species Mouse
Isotype κ
Immunogen Microsomes from A431 cells
Antigen EGF R/ErbB1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunoprecipitation
Test Specificity Human. Does not react with Mouse or Rat.
Gene Accession No. P00533
Gene ID (Entrez) 1956
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL