Please login to your online account to display your discounted pricing

anti-Dynamin, Clone: 3G4B6, Novus Biologicals™

$139.00 - $309.05

Specifications

Clone 3G4B6
Host Species Murine
Isotype IgG2a
Immunogen Purified recombinant fragment of human Dynamin expressed in &italic_begin;E. coli&italic_end;. [UniProt No. Q05193]
Antigen Dynamin
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11060491

 
novus biologicals canada
NB11060491
Unlabeled 0.1mL Each for $309.05

NB11060491T

 
novus biologicals canada
NB11060491SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Clone 3G4B6
Host Species Murine
Isotype IgG2a
Immunogen Purified recombinant fragment of human Dynamin expressed in &italic_begin;E. coli&italic_end;. [UniProt No. Q05193]
Antigen Dynamin
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Gene Accession No. Q05193
Gene ID (Entrez) 1759