Please login to your online account to display your discounted pricing

anti-DNMT3B, Clone: 52A1018, Novus Biologicals™

$144.63 - $384.00

Specifications

Clone 52A1018
Host Species Murine
Isotype κ
Immunogen This antibody was raised against bacteria expressed HIS-tag recombinant full length mouse Dnmt3b. The antibody will also recognize human Dnmt3b.
Antigen DNMT3B
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056514

 
novus biologicals canada
NB10056514
Unlabeled 0.1mg Each for $384.00

NB10056514T

 
novus biologicals canada
NB10056514SS
Unlabeled 0.025mg Each for $144.63
Description & Specifications

Specifications

Clone 52A1018
Host Species Murine
Isotype κ
Immunogen This antibody was raised against bacteria expressed HIS-tag recombinant full length mouse Dnmt3b. The antibody will also recognize human Dnmt3b.
Antigen DNMT3B
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Chromatin Immunoprecipitation
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blot
Test Specificity This does not react with Dnmt3a. Reacts with Human and Mouse. Not yet tested in other species.
Gene Accession No. Q9UBC3
Gene ID (Entrez) 1789
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL