Please login to your online account to display your discounted pricing

anti-DNMT3B, Clone: 52A1018, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056514

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056514

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 1789
Gene Accession No. Q9UBC3
Primary or Secondary Primary
Conjugate Unlabeled
Formulation PBS containing 0.05% BSA
Monoclonal or Polyclonal Monoclonal
Test Specificity This does not react with Dnmt3a. Reacts with Human and Mouse. Not yet tested in other species.
Concentration 0.5mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Applications Chromatin Immunoprecipitation
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Isotype κ
Quantity 0.1mg
Format Purified
Host Species Mouse
Antigen DNMT3B
Clone 52A1018
Regulatory Status RUO
Immunogen This antibody was raised against bacteria expressed HIS-tag recombinant full length mouse Dnmt3b. The antibody will also recognize human Dnmt3b.