Please login to your online account to display your discounted pricing

anti-Dnmt2, Clone: 102B1259.2, Novus Biologicals™


Manufacturer: novus biologicals canada  NB200587SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB200587SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 1787
Monoclonal or Polyclonal Monoclonal
Test Specificity In mouse Dnmt2 transfected cell lysate, a 42 kDa band is oberseved. Cross-reacts with Mouse. Not yet tested in other species.
Conjugate Unlabeled
Formulation PBS containing 0.2% gelatin
Primary or Secondary Primary
Gene Accession No. Q6ICS7
Immunogen This antibody was raised against synthetic peptides corresponding to mouse Dnmt2.
Clone 102B1259.2
Concentration 0.5mg/mL
Format Purified
Applications Western Blotting
Isotype κ
Cross Reactivity Mouse
Antigen Dnmt2
Host Species Mouse
Quantity 0.025mg
Regulatory Status RUO