Please login to your online account to display your discounted pricing

anti-DMC1, Clone: 1D12/4, Novus Biologicals™

$139.00 - $309.05

Specifications

Clone 1D12/4
Host Species Mouse
Isotype IgG1
Immunogen His-tagged human DMC1
Antigen DMC1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100181

 
novus biologicals canada
NB100181
Unlabeled 0.1mL Each for $309.05

NB100181SS

 
novus biologicals canada
NB100181SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Clone 1D12/4
Host Species Mouse
Isotype IgG1
Immunogen His-tagged human DMC1
Antigen DMC1
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Human and mouse. Does not cross-react with human Rad51 in Western analysis.
Gene Accession No. Q14565
Gene ID (Entrez) 11144