Please login to your online account to display your discounted pricing

anti-Desmoglein-3, Clone: 5H10, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 5H10
Host Species Murine
Isotype IgG1
Immunogen The extracellular domain of human Desmoglein 3 [Swiss-Prot No. P32926]
Antigen Desmoglein-3
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP178984

 
novus biologicals canada
NBP178984
Unlabeled 0.1mL Each for $309.05

NBP178984SS

 
novus biologicals canada
NBP178984SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Clone 5H10
Host Species Murine
Isotype IgG1
Immunogen The extracellular domain of human Desmoglein 3 [Swiss-Prot No. P32926]
Antigen Desmoglein-3
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Gene Accession No. P32926
Gene ID (Entrez) 1830
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL