missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-DDX3, Clone: 11F11D10, Novus Biologicals™

$139.00 - $255.80

Specifications

Clone 11F11D10
Host Species Murine
Isotype κ
Immunogen A partial recombinant protein containing amino acids 24-203 of human DDX3 was used as immunogen for this antibody.
Antigen DDX3
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
NBP214848 Novus Biologicals Canada
NBP214848
0.1mg Each for $255.80
NBP214848SS Novus Biologicals Canada
NBP214848SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

11F11D10
κ
DDX3
Purified
RUO
O00571
PBS containing 0.05% BSA
Murine
A partial recombinant protein containing amino acids 24-203 of human DDX3 was used as immunogen for this antibody.
Monoclonal
Primary
Flow Cytometry,Western Blot
1654
0.5mg/mL