Please login to your online account to display your discounted pricing

anti-DC-SIGN/CD209, Clone: 111H2.02, Novus Biologicals™

$309.05 - $586.39

Specifications

Clone 111H2.02
Host Species Mouse
Isotype IgG2b
Immunogen HeLa cells stably transfected-with human DC-SIGN
Antigen DC-SIGN/CD209
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

DDX0209

 
novus biologicals canada
DDX0209
Unlabeled 0.1mg Each for $586.39

DDX0209005

 
novus biologicals canada
DDX0209005MG
Unlabeled 0.05mg Each for $309.05
Description & Specifications

Specifications

Clone 111H2.02
Host Species Mouse
Isotype IgG2b
Immunogen HeLa cells stably transfected-with human DC-SIGN
Antigen DC-SIGN/CD209
Monoclonal or Polyclonal Monoclonal
Format Ion Exchange Chromatography
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry
Gene Accession No. NP_066978
Gene ID (Entrez) 30835
Formulation Antibody in Tris-NaCl pH 8