Please login to your online account to display your discounted pricing

anti-DC-SIGN/CD209, Clone: 111H2.02, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX0209005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX0209005

  • / Each
 

Description & Specifications

Specifications

Antigen DC-SIGN/CD209
Applications Flow Cytometry
Applications Immunohistochemistry
Clone 111H2.02
Conjugate Unlabeled
Cross Reactivity Human
Format Ion Exchange Chromatography
Formulation Antibody in Tris-NaCl pH 8
Gene Accession No. NP_066978
Host Species Murine
Immunogen HeLa cells stably transfected-with human DC-SIGN
Isotype IgG2b
Quantity 0.05mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 30835
Target Species Human