Please login to your online account to display your discounted pricing

anti-DC-SIGN/CD209, Clone: 109H12.03, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX0207005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX0207005

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 30835
Antigen DC-SIGN/CD209
Immunogen human DC-SIGN-transfected HeLa cells
Quantity 0.05mg
Format Ion Exchange Chromatography
Host Species Mouse
Applications Flow Cytometry
Applications Immunohistochemistry
Isotype IgG2a
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Clone 109H12.03
Formulation Antibody in Tris-NaCl pH 8
Gene Accession No. NP_066978
Primary or Secondary Primary
Conjugate Unlabeled
Cross Reactivity Human