Please login to your online account to display your discounted pricing

anti-DC-SIGN/CD209, Clone: 109H12.03, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX0207

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX0207

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 30835
Cross Reactivity Human
Regulatory Status RUO
Quantity 0.1mg
Clone 109H12.03
Applications Flow Cytometry
Applications Immunohistochemistry
Immunogen human DC-SIGN-transfected HeLa cells
Isotype IgG2a
Antigen DC-SIGN/CD209
Formulation Antibody in Tris-NaCl pH 8
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Gene Accession No. NP_066978
Conjugate Unlabeled
Host Species Mouse
Format Ion Exchange Chromatography