Please login to your online account to display your discounted pricing

anti-DC-SIGN/CD209, Biotin, Clone: 111H2.02, Novus Biologicals™

$406.00 - $536.00

Specifications

Clone 111H2.02
Host Species Mouse
Isotype IgG2b
Immunogen HeLa cells stably transfected-with human DC-SIGN
Antigen DC-SIGN/CD209
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

DDX0209B005

 
novus biologicals canada
DDX0209B005MG
Biotin 0.05mg Each for $406.00

DDX0209B

 
novus biologicals canada
DDX0209B
Biotin 0.1mg Each for $536.00
Description & Specifications

Specifications

Clone 111H2.02
Host Species Mouse
Isotype IgG2b
Immunogen HeLa cells stably transfected-with human DC-SIGN
Antigen DC-SIGN/CD209
Monoclonal or Polyclonal Monoclonal
Format Ion Exchange Chromatography
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Gene Accession No. NP_066978
Gene ID (Entrez) 30835
Formulation Antibody in PBS 50% glycerol