Please login to your online account to display your discounted pricing

anti-DC-SIGN/CD209, Biotin, Clone: 111H2.02, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX0209B005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX0209B005

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 30835
Cross Reactivity Human
Immunogen HeLa cells stably transfected-with human DC-SIGN
Applications Immunohistochemistry
Isotype IgG2b
Antigen DC-SIGN/CD209
Format Ion Exchange Chromatography
Regulatory Status RUO
Clone 111H2.02
Gene Accession No. NP_066978
Monoclonal or Polyclonal Monoclonal
Conjugate Biotin
Primary or Secondary Primary
Formulation Antibody in PBS 50% glycerol
Quantity 0.05mg
Host Species Mouse