Please login to your online account to display your discounted pricing

anti-DC-SIGN/CD209, Biotin, Clone: 109H12.03, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX0207B

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX0207B

  • / Each
 

Description & Specifications

Specifications

Antigen DC-SIGN/CD209
Applications Flow Cytometry
Applications Immunohistochemistry
Clone 109H12.03
Conjugate Biotin
Cross Reactivity Human
Format Ion Exchange Chromatography
Formulation Antibody in PBS 50% glycerol
Gene Accession No. NP_066978
Host Species Murine
Immunogen human DC-SIGN-transfected HeLa cells
Isotype IgG2a
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 30835
Target Species Human