Please login to your online account to display your discounted pricing

anti-DC-SIGN/CD209, Biotin, Clone: 109H12.03, Novus Biologicals™

$406.00 - $536.00

Specifications

Clone 109H12.03
Host Species Murine
Isotype IgG2a
Immunogen human DC-SIGN-transfected HeLa cells
Antigen DC-SIGN/CD209
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

DDX0207B

 
novus biologicals canada
DDX0207B
Biotin 0.1mg Each for $536.00

DDX0207B005

 
novus biologicals canada
DDX0207B005MG
Biotin 0.05mg Each for $406.00
Description & Specifications

Specifications

Clone 109H12.03
Host Species Murine
Isotype IgG2a
Immunogen human DC-SIGN-transfected HeLa cells
Antigen DC-SIGN/CD209
Monoclonal or Polyclonal Monoclonal
Format Ion Exchange Chromatography
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry
Gene Accession No. NP_066978
Gene ID (Entrez) 30835
Formulation Antibody in PBS 50% glycerol