Please login to your online account to display your discounted pricing

anti-DC-SIGN/CD209, Biotin, Clone: 109H12.03, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX0207B005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX0207B005

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 30835
Applications Flow Cytometry
Applications Immunohistochemistry
Immunogen human DC-SIGN-transfected HeLa cells
Antigen DC-SIGN/CD209
Format Ion Exchange Chromatography
Host Species Mouse
Isotype IgG2a
Clone 109H12.03
Cross Reactivity Human
Formulation Antibody in PBS 50% glycerol
Gene Accession No. NP_066978
Quantity 0.05mg
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Biotin
Regulatory Status RUO