Please login to your online account to display your discounted pricing

anti-DAP5, Clone: 39C534.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB12011929

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB12011929

  • / Each
 

Description & Specifications

Specifications

Antigen DAP5
Applications Flow Cytometry
Applications Western Blot
Clone 39C534.1
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation PBS containing 0.2% gelatin
Gene Accession No. P78344
Host Species Murine
Immunogen The antibody was raised against two synthetic peptides corresponding to amino acids 796-812 of human DAP-5 protein.
Isotype IgG1
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1982
Test Specificity In Jurkat, a 97 kDa band is observed.
Target Species Human
Target Species Murine