Please login to your online account to display your discounted pricing

anti-DAP5, Clone: 39C534.1, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 39C534.1
Host Species Murine
Isotype IgG1
Immunogen The antibody was raised against two synthetic peptides corresponding to amino acids 796-812 of human DAP-5 protein.
Antigen DAP5
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB12011929T

 
novus biologicals canada
NB12011929SS
Unlabeled 0.025mg Each for $109.00

NB12011929

 
novus biologicals canada
NB12011929
Unlabeled 0.1mg Each for $309.05
Description & Specifications

Specifications

Clone 39C534.1
Host Species Murine
Isotype IgG1
Immunogen The antibody was raised against two synthetic peptides corresponding to amino acids 796-812 of human DAP-5 protein.
Antigen DAP5
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Western Blot
Test Specificity In Jurkat, a 97 kDa band is observed.
Gene Accession No. P78344
Gene ID (Entrez) 1982
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL