Please login to your online account to display your discounted pricing

anti-DAP5, Clone: 39C534.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB12011929SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB12011929T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 1982
Gene Accession No. P78344
Test Specificity In Jurkat, a 97 kDa band is observed.
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL
Conjugate Unlabeled
Host Species Mouse
Applications Flow Cytometry
Applications Western Blotting
Isotype IgG1
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Immunogen The antibody was raised against two synthetic peptides corresponding to amino acids 796-812 of human DAP-5 protein.
Cross Reactivity Human
Cross Reactivity Mouse
Antigen DAP5
Regulatory Status RUO
Clone 39C534.1
Format Purified
Quantity 0.025mg