Please login to your online account to display your discounted pricing

anti-DAP3, Clone: 42C617.1.2, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 42C617.1.2
Host Species Murine
Isotype IgG
Immunogen The antibody was raised against a synthetic peptide corresponding to amino acids 51-66 of human DAP-3 protein.
Antigen DAP3
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056539

 
novus biologicals canada
NB10056539
Unlabeled 0.1mg Each for $329.00

NB10056539T

 
novus biologicals canada
NB10056539SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 42C617.1.2
Host Species Murine
Isotype IgG
Immunogen The antibody was raised against a synthetic peptide corresponding to amino acids 51-66 of human DAP-3 protein.
Antigen DAP3
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity In Jurkat, a 45 kDa band is observed. Reacts with Mouse and Human. 75% sequence identity with rat protein.
Gene Accession No. P51398
Gene ID (Entrez) 7818
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL