Please login to your online account to display your discounted pricing

anti-D4-GDI/RhoGDI2, Clone: 10D774, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056556

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056556

  • / Each
 

Description & Specifications

Specifications

Antigen D4-GDI/RhoGDI2
Applications Flow Cytometry
Applications Western Blot
Clone 10D774
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS containing 0.02% gelatin.
Host Species Murine
Immunogen A synthetic peptide corresponding to human D4-GDI/RhoGDI2 was used as the immunogen.
Isotype IgG
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 397
Test Specificity A 28 kDa band is observed.
Target Species Human