Please login to your online account to display your discounted pricing

anti-D4-GDI/RhoGDI2, Clone: 10D774, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 10D774
Host Species Murine
Isotype IgG
Immunogen A synthetic peptide corresponding to human D4-GDI/RhoGDI2 was used as the immunogen.
Antigen D4-GDI/RhoGDI2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056556

 
novus biologicals canada
NB10056556
Unlabeled 0.1mg Each for $329.00

NB10056556T

 
novus biologicals canada
NB10056556SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 10D774
Host Species Murine
Isotype IgG
Immunogen A synthetic peptide corresponding to human D4-GDI/RhoGDI2 was used as the immunogen.
Antigen D4-GDI/RhoGDI2
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Western Blot
Test Specificity A 28 kDa band is observed.
Gene ID (Entrez) 397
Formulation PBS containing 0.02% gelatin.