Please login to your online account to display your discounted pricing

anti-D4-GDI/RhoGDI2, Clone: 10D774, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056556SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056556T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 397
Monoclonal or Polyclonal Monoclonal
Test Specificity A 28 kDa band is observed.
Formulation PBS containing 0.02% gelatin.
Primary or Secondary Primary
Conjugate Unlabeled
Antigen D4-GDI/RhoGDI2
Format Purified
Immunogen A synthetic peptide corresponding to human D4-GDI/RhoGDI2 was used as the immunogen.
Quantity 0.025mg
Cross Reactivity Human
Applications Flow Cytometry
Applications Western Blotting
Regulatory Status RUO
Host Species Mouse
Isotype IgG
Clone 10D774