Please login to your online account to display your discounted pricing

anti-Cytokeratin, HMW, Clone: 34bE12, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone 34bE12
Host Species Mouse
Isotype IgG1
Immunogen Solublized keratin extract from human stratum corneum was used as the immunogen for the Cytokeratin 34bE12 antibody.
Antigen Cytokeratin
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP229417

 
novus biologicals canada
NBP229417
Unlabeled 0.5mL Each for $321.00

NBP2294171M

 
novus biologicals canada
NBP2294171ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone 34bE12
Host Species Mouse
Isotype IgG1
Immunogen Solublized keratin extract from human stratum corneum was used as the immunogen for the Cytokeratin 34bE12 antibody.
Antigen Cytokeratin
Monoclonal or Polyclonal Monoclonal
Format Concentrated
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity This antibody recognizes CK1, CK5, CK10 and CK14. In normal epithelia, it stains stratified epithelia, myoepithelial cells and basal cells in the prostate gland and bronchi. This MAb shows no reactivity with hepatocytes, pancreatic acinar cells, proximal renal tubules, or endometrial glands; there is no reactivity with cells derived from simple epithelia. Mesenchymal tumors, lymphomas, melanomas, neural tumors, and neuroendocrine tumors are negative with this antibody. It stains myoepithelial cells and has been shown to be useful in distinguishing prostate adenocarcinoma from benign prostate. This antibody has also been useful in separating benign from malignant intraductal breast proliferations.
Gene Accession No. P13646
Gene ID (Entrez) 51350
Formulation Culture Medium