Please login to your online account to display your discounted pricing

anti-Cytokeratin 4, Clone: 6B10, Novus Biologicals™

$332.00 - $1,181.00

Specifications

Clone 6B10
Host Species Mouse
Isotype IgG1
Immunogen Cytokeratin preparation purified form Human esophagus
Antigen Cytokeratin 4
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP1421361M

 
novus biologicals canada
NBP1421361ML
Unlabeled 1mL Each for $332.00

NBP1421365M

 
novus biologicals canada
NBP1421365ML
Unlabeled 5mL Each for $1,181.00
Description & Specifications

Specifications

Clone 6B10
Host Species Mouse
Isotype IgG1
Immunogen Cytokeratin preparation purified form Human esophagus
Antigen Cytokeratin 4
Monoclonal or Polyclonal Monoclonal
Format Supernatant
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Test Specificity The antibody does stain non-cornifying squamous epithelium, cells in certain ciliated pseudostratified epithelia and ductal epithelia of various exocrine glands. In immunoblotting experiments the antibody recognizes only keratin no. 4 (59kD) and does not show any cross reactivity with other cytokeratins. Immunogen: Cytokeratin preparation purified form Human esophagus.
Gene ID (Entrez) 3851