Please login to your online account to display your discounted pricing

anti-Cytokeratin 4, Clone: 6B10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1421365ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1421365M

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Rat
Isotype IgG1
Antigen Cytokeratin 4
Format Supernatant
Host Species Mouse
Clone 6B10
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Immunogen Cytokeratin preparation purified form Human esophagus
Gene ID (Entrez) 3851
Test Specificity The antibody does stain non-cornifying squamous epithelium, cells in certain ciliated pseudostratified epithelia and ductal epithelia of various exocrine glands. In immunoblotting experiments the antibody recognizes only keratin no. 4 (59kD) and does not show any cross reactivity with other cytokeratins. Immunogen: Cytokeratin preparation purified form Human esophagus.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Quantity 5mL
Regulatory Status RUO