Please login to your online account to display your discounted pricing

anti-Cytokeratin 4, Clone: 6B10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1421361ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1421361M

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Antigen Cytokeratin 4
Host Species Mouse
Isotype IgG1
Regulatory Status RUO
Format Supernatant
Immunogen Cytokeratin preparation purified form Human esophagus
Clone 6B10
Cross Reactivity Human
Cross Reactivity Rat
Quantity 1mL
Primary or Secondary Primary
Gene ID (Entrez) 3851
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Test Specificity The antibody does stain non-cornifying squamous epithelium, cells in certain ciliated pseudostratified epithelia and ductal epithelia of various exocrine glands. In immunoblotting experiments the antibody recognizes only keratin no. 4 (59kD) and does not show any cross reactivity with other cytokeratins. Immunogen: Cytokeratin preparation purified form Human esophagus.