Please login to your online account to display your discounted pricing

anti-Cytokeratin 10, Clone: LH2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329611ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329611M

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1
Format Purified
Quantity 1mL
Cross Reactivity Human
Cross Reactivity Mouse
Host Species Mouse
Immunogen Skin extract of a human Psoriasis patient
Clone LH2
Antigen Cytokeratin 10
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Gene Accession No. P13645
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3858
Concentration 0.2mg/mL