Please login to your online account to display your discounted pricing

anti-Cytokeratin 10, Clone: LH2, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone LH2
Host Species Mouse
Isotype IgG1
Immunogen Skin extract of a human Psoriasis patient
Antigen Cytokeratin 10
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2329611M

 
novus biologicals canada
NBP2329611ML
Unlabeled 1mL Each for $429.00

NBP232961

 
novus biologicals canada
NBP232961
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone LH2
Host Species Mouse
Isotype IgG1
Immunogen Skin extract of a human Psoriasis patient
Antigen Cytokeratin 10
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P13645
Gene ID (Entrez) 3858
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL