Please login to your online account to display your discounted pricing

anti-Cytokeratin 1, Clone: LHK1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002756

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002756

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 3848
Concentration 1.14mg/mL
Formulation Tris-glycine, 150mM NaCl
Gene Accession No. P04264
Primary or Secondary Primary
Conjugate Unlabeled
Host Species Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Immunogen C-terminal peptide sequence of human Cytokeratin 1 [UniProt No. P04264]
Clone LHK1
Isotype κ
Cross Reactivity Human
Cross Reactivity Rat
Format Purified
Regulatory Status RUO
Antigen Cytokeratin 1
Quantity 0.1mL