Please login to your online account to display your discounted pricing

anti-Cytochrome c, Clone: 7H8.2C12, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056503

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056503

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Gene Accession No. P99999
Formulation PBS containing 0.5% BSA
Gene ID (Entrez) 54205
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Test Specificity An approx. 15 kDa band is observed. This antibody recognizes total cytochrome C which includes both apocytochrome (i.e. cytochrome in the cytosol without heme attached) and holocytochrome (i.e cytochrome in the mitochondria with heme attached). Cross reacts with Canine, Drosophila, Honey Bee, Equine, Human, Mammals, Porcine and Rabbit. Mouse reactivity reported in multiple pieces of scientific literature.
Conjugate Unlabeled
Quantity 0.1mg
Regulatory Status RUO
Cross Reactivity Canine
Cross Reactivity Drosophila
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mammalia
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rabbit
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Isotype κ
Immunogen 81-104 and 66-104 of pigeon CYT were used as the immunogen (Jemmerson et al. 1991). The antibody recognizes an epitope within amino acids 93-104 of pigeon cytochrome C based on competitive ELISA results (Jemmerson et al. 1991).
Format Purified
Antigen Cytochrome c
Host Species Mouse
Clone 7H8.2C12