Please login to your online account to display your discounted pricing

anti-Cytochrome c, Clone: 7H8.2C12, Novus Biologicals™

$139.00 - $362.00

Specifications

Clone 7H8.2C12
Host Species Murine
Isotype κ
Immunogen 81-104 and 66-104 of pigeon CYT were used as the immunogen (Jemmerson et al. 1991). The antibody recognizes an epitope within amino acids 93-104 of pigeon cytochrome C based on competitive ELISA results (Jemmerson et al. 1991).
Antigen Cytochrome c
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056503

 
novus biologicals canada
NB10056503
Unlabeled 0.1mg Each for $362.00

NB10056503T

 
novus biologicals canada
NB10056503SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 7H8.2C12
Host Species Murine
Isotype κ
Immunogen 81-104 and 66-104 of pigeon CYT were used as the immunogen (Jemmerson et al. 1991). The antibody recognizes an epitope within amino acids 93-104 of pigeon cytochrome C based on competitive ELISA results (Jemmerson et al. 1991).
Antigen Cytochrome c
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity An approx. 15 kDa band is observed. This antibody recognizes total cytochrome C which includes both apocytochrome (i.e. cytochrome in the cytosol without heme attached) and holocytochrome (i.e cytochrome in the mitochondria with heme attached). Cross reacts with Canine, Drosophila, Honey Bee, Equine, Human, Mammals, Porcine and Rabbit. Mouse reactivity reported in multiple pieces of scientific literature.
Gene Accession No. P99999
Gene ID (Entrez) 54205
Formulation PBS containing 0.5% BSA
Concentration 0.5mg/mL