Please login to your online account to display your discounted pricing

anti-CTLA-4, Clone: 37B3D8.2F2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP231143SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP231143SS

  • / Each
 

Description & Specifications

Specifications

Format Purified
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1493
Cross Reactivity Human
Cross Reactivity Mouse
Quantity 0.025mg
Immunogen mouse CTLA4
Concentration 1.0mg/mL
Regulatory Status RUO
Host Species Mouse
Applications Western Blotting
Antigen CTLA-4
Clone 37B3D8.2F2
Primary or Secondary Primary
Conjugate Unlabeled