Please login to your online account to display your discounted pricing

anti-CTLA-4, Clone: 37B3D8.2F2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP231143

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP231143

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Conjugate Unlabeled
Antigen CTLA-4
Immunogen mouse CTLA4
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Quantity 0.1mg
Host Species Mouse
Applications Western Blotting
Regulatory Status RUO
Gene ID (Entrez) 1493
Clone 37B3D8.2F2
Concentration 1.0mg/mL
Monoclonal or Polyclonal Monoclonal