Please login to your online account to display your discounted pricing

anti-CREB, Clone: 82B514.2, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056525SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056525T

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Formulation PBS containing 0.2% gelatin
Primary or Secondary Primary
Test Specificity In HeLa, a 43 kDa band is observed.
Gene Accession No. NP_604391
Gene ID (Entrez) 1385
Antigen CREB
Format Purified
Host Species Mouse
Applications Western Blotting
Regulatory Status RUO
Concentration 0.5mg/mL
Cross Reactivity Human
Immunogen A portion of amino acids 100-150 of human CREB was used as the immunogen.
Clone 82B514.2
Isotype IgG1
Quantity 0.025mg