Please login to your online account to display your discounted pricing

anti-CREB, Clone: 82B514.2, Novus Biologicals™

$139.00 - $259.02

Specifications

Clone 82B514.2
Host Species Murine
Isotype IgG1
Immunogen A portion of amino acids 100-150 of human CREB was used as the immunogen.
Antigen CREB
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056525

 
novus biologicals canada
NB10056525
Unlabeled 0.1mg Each for $259.02

NB10056525T

 
novus biologicals canada
NB10056525SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 82B514.2
Host Species Murine
Isotype IgG1
Immunogen A portion of amino acids 100-150 of human CREB was used as the immunogen.
Antigen CREB
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity In HeLa, a 43 kDa band is observed.
Gene Accession No. NP_604391
Gene ID (Entrez) 1385
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL