Please login to your online account to display your discounted pricing

anti-&italic_begin;Clostridium difficile&italic_end; Toxin A, Clone: PCG4.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6001066

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6001066

  • / Each
 

Description & Specifications

Specifications

Test Specificity C. difficile Toxin A. Does not cross react with C. difficile Toxin B. Cross-reacts with C. difficile. Does not cross react with C. difficile toxin B.
Isotype κ
Applications ELISA
Applications Western Blotting
Regulatory Status RUO
Format Purified
Immunogen Full length protein (&italic_begin;C. difficile&italic_end;).
Quantity 1mg
Formulation 10mM Sodium Phosphate, 7.4 containing 150mM Sodium Chloride
Concentration 1.16mg/mL
Primary or Secondary Primary
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Bacteria
Antigen Clostridium difficile Toxin A
Host Species Mouse
Clone PCG4.1